Boc-L-β-homoisoleucine ≥98%

Supplier: Ambeed
Warning

Synonyms: Boc-β-HomoIle-OH, Boc-β-HoIle-OH

A270748-5G A270748-1G A270748-250MG
77593-672EA 751.33 USD
77593-672 77626-404 77626-406
Boc-L-β-homoisoleucine ≥98%
Boc-L-beta-homoisoleucine

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₄
MW: 245.32 g/mol
MDL Number: MFCD01076254
CAS Number: 218608-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR