N,N-Dimethylimidodicarbonimidic diamide ≥95%

Supplier: Ambeed
Warning

Synonyms: Metformin

A270611-1G A270611-250MG A270611-10G
77616-142EA 18.54 USD
77616-142 77616-144 77746-476
N,N-Dimethylimidodicarbonimidic diamide ≥95%
N,N-Dimethylimidodicarbonimidic diamide

Precaución: for Research Use Only

Formula: C₄H₁₁N₅
MW: 129.16 g/mol
MDL Number: MFCD00242652
CAS Number: 657-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR