5-Bromo-2-(trifluoromethoxy)phenol ≥98%

Supplier: Ambeed
Warning

A270361-25G A270361-5G
77612-584EA 266.77 USD
77612-584 77683-823
5-Bromo-2-(trifluoromethoxy)phenol ≥98%
5-Bromo-2-(trifluoromethoxy)phenol

Precaución: For research use only.

Formula: C₇H₄BrF₃O₂
MW: 257.005 g/mol
MDL Number: MFCD21333031
CAS Number: 1048963-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR