6-(3-Fluorophenyl)nicotinic acid ≥95%

Supplier: Ambeed
Warning

A270150-1G A270150-250MG
77872-314EA 108 USD
77872-314 77872-315
6-(3-Fluorophenyl)nicotinic acid ≥95%
6-(3-Fluorophenyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₂H₈FNO₂
MDL Number: MFCD06410387
CAS Number: 582325-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR