1-Bromo-3-cyclopropylbenzene ≥98%

Supplier: Ambeed
Warning

A269866-1G A269866-250MG A269866-5G
77746-472EA 50 USD
77746-472 77746-473 77746-474
1-Bromo-3-cyclopropylbenzene ≥98%
1-Bromo-3-cyclopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₉Br
MDL Number: MFCD01070823
CAS Number: 1798-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR