Amberlite® IR-120 (H) moisture 53-58%

Supplier: Ambeed
Warning

A269865-25G A269865-500G
77877-901EA 12 USD
77877-901 77877-902
Amberlite® IR-120 (H) moisture 53-58%
Amberlite® IR-120 (H)

Precaución: for Research Use Only

MDL Number: MFCD00132707
CAS Number: 78922-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR