Aztreonam ≥98%

Supplier: Ambeed
Danger

Synonyms: 2-[[(Z)-[1-(2-Amino-4-thiazolyl)-2-[[(2S,3S)-2-methyl-4-oxo-1-sulpho-3-azetidinyl]amino]-2-oxoethylidene]amino]oxy]-2-methylpropionic Acid, 2-((((Z)-1-(2-Aminothiazol-4-yl)-2-(((2S,3S)-2-methyl-4-oxo-1-sulphoazetidin-3-yl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic acid

A269798-1G A269798-1MG A269798-250MG A269798-5G
77612-576EA 66.95 USD
77612-576 77672-392 77672-394 77672-396
Aztreonam ≥98%
Aztreonam

Precaución: for Research Use Only

Formula: C₁₃H₁₇N₅O₈S₂
MW: 435.43 g/mol
MDL Number: MFCD00072145
CAS Number: 78110-38-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR