(4-Benzylphenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A269737-100MG A269737-1G A269737-250MG A269737-5G
77885-708EA 55 USD
77885-708 77885-709 77885-710 77885-711
(4-Benzylphenyl)boronic acid ≥98%
(4-Benzylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃BO₂
MDL Number: MFCD08729295
CAS Number: 56311-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR