Nickel(II) dibutyldithiocarbamate ≥97%

Supplier: Ambeed
Danger

Synonyms: Nickel dibutyldithiocarbamate, Nickel(II) Bis(dibutyldithiocarbamate), Dibutyldithiocarbamic Acid Nickel(II) Salt

A269680-100G A269680-500G
77601-156EA 15.38 USD
77601-156 77672-390
Nickel(II) dibutyldithiocarbamate ≥97%
Nickel(II) dibutyldithiocarbamate

Precaución: for Research Use Only

Formula: C₉H₁₉NS₂
MDL Number: MFCD00067273
CAS Number: 13927-77-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR