5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine ≥96%

Supplier: Ambeed
Warning

Synonyms: Ticlopidine

A269629-250MG A269629-1G A269629-100MG
77593-668EA 92.25 USD
77593-668 77601-154 77668-176
5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine ≥96%
5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₄ClNS
MW: 263.78 g/mol
MDL Number: MFCD00661081
CAS Number: 55142-85-3
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR