GSK-484 hydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: GSK484 HCl, ((3S,4R)-3-Amino-4-hydroxypiperidin-1-yl)(2-(1-(cyclopropylmethyl)-1H-indol-2-yl)-7-methoxy-1-methyl-1H-benzo[d]imidazol-5-yl)methanone hydrochloride

A268467-100MG A268467-50MG A268467-250MG
77622-278EA 494.4 USD
77622-278 77622-280 77783-377
GSK-484 hydrochloride ≥98%
GSK-484 hydrochloride

Precaución: for Research Use Only

Formula: C₂₇H₃₂ClN₅O₃
MDL Number: MFCD30343874
CAS Number: 1652591-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR