tert-Butyl-4-(bromomethyl)-1,3-thiazol-2-ylcarbamate ≥97%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl (4-(bromomethyl)thiazol-2-yl)carbamate

A268438-100MG A268438-250MG
77616-134EA 31.93 USD
77616-134 77746-465
tert-Butyl-4-(bromomethyl)-1,3-thiazol-2-ylcarbamate ≥97%
tert-Butyl-4-(bromomethyl)-1,3-thiazol-2-ylcarbamate

Precaución: for Research Use Only

Formula: C₉H₁₃BrN₂O₂S
MW: 293.19 g/mol
MDL Number: MFCD09751127
CAS Number: 1001419-35-7
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR