Ethyl 2-bromo-1H-imidazole-5-carboxylate ≥98%

Supplier: Ambeed
Warning

A268250-100MG A268250-250MG
77593-660EA 62.53 USD
77593-660 77885-705
Ethyl 2-bromo-1H-imidazole-5-carboxylate ≥98%
Ethyl 2-bromo-1H-imidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
MDL Number: MFCD10697827
CAS Number: 74478-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR