1'-Hydroxy-[1,1'-bi(cyclohexan)]-2-one ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A268212-1G A268212-100MG
77593-658EA 306.48 USD
77593-658 77683-820
1'-Hydroxy-[1,1'-bi(cyclohexan)]-2-one ≥95%, Ambeed.Inc
1'-Hydroxy-[1,1'-bi(cyclohexan)]-2-one

Precaución: For research use only.

Formula: C₁₂H₂₀O₂
MW: 196.29 g/mol
MDL Number: MFCD03837683
CAS Number: 28746-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR