Palladium, [1,3-bis[2,6-bis(1-methylethyl)phenyl]-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(1-methyl-1H-imidazole-kN3)-, (SP-4-1)- ≥97%

Supplier: Ambeed
Warning

A267026-100MG A267026-1G A267026-250MG
77612-560EA 46.35 USD
77612-560 77612-562 77612-564
Palladium, [1,3-bis[2,6-bis(1-methylethyl)phenyl]-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(1-methyl-1H-imidazole-kN3)-, (SP-4-1)- ≥97%
Palladium, [1,3-bis[2,6-bis(1-methylethyl)phenyl]-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(1-methyl-1H-imidazole-kN3)-, (SP-4-1)-

Precaución: for Research Use Only

Formula: C₃₁H₄₂Cl₂N₄Pd
MW: 648.02 g/mol
MDL Number: MFCD28252260
CAS Number: 1314876-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR