6-(3-Amino-1H-indazol-4-yl)-N-(4-fluoro-3-methylphenyl)-1-naphthamide ≥98%

Supplier: Ambeed
Warning

Synonyms: SYHA1813

A2668735-1MG A2668735-10MG A2668735-5MG
77598-380EA 418.2 USD
77598-380 77626-394 77626-396
6-(3-Amino-1H-indazol-4-yl)-N-(4-fluoro-3-methylphenyl)-1-naphthamide ≥98%
6-(3-Amino-1H-indazol-4-yl)-N-(4-fluoro-3-methylphenyl)-1-naphthamide

Precaución: for Research Use Only

Formula: C₂₅H₁₉FN₄O
MW: 410.44 g/mol
CAS Number: 1807466-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR