5-(((1R,1aS,6bR)-1-(6-(Trifluoromethyl)-1H-benzo[d]imidazol-2-yl)-1a,6b-dihydro-1H-cyclopropa[b]benzofuran-5-yl)oxy)-3,4-dihydro-1,8-naphthyridin-2(1H)-one ≥96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Lifirafenib
A266866-1MG
A266866-5MG
77746-459EA
56
USD
77746-459
77746-460
5-(((1R,1aS,6bR)-1-(6-(Trifluoromethyl)-1H-benzo[d]imidazol-2-yl)-1a,6b-dihydro-1H-cyclopropa[b]benzofuran-5-yl)oxy)-3,4-dihydro-1,8-naphthyridin-2(1H)-one ≥96%
5-(((1R,1aS,6bR)-1-(6-(Trifluoromethyl)-1H-benzo[d]imidazol-2-yl)-1a,6b-dihydro-1H-cyclopropa[b]benzofuran-5-yl)oxy)-3,4-dihydro-1,8-naphthyridin-2(1H)-one
Precaución: for Research Use Only
Formula:
C₂₅H₁₇F₃N₄O₃ MW: 478.42 g/mol |
MDL Number:
MFCD30489379 CAS Number: 1446090-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...