Ertapenem sodium salt ≥97% (by HPLC)

Supplier: Ambeed
Warning

A266766-10MG A266766-1MG A266766-50MG
77612-554EA 14.42 USD
77612-554 77612-556 77683-817
Ertapenem sodium salt ≥97% (by HPLC)
Ertapenem sodium salt

Precaución: For research use only.

Formula: C₂₂H₂₄N₃NaO₇S
MDL Number: MFCD09475566
CAS Number: 153773-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR