Ac-DL-β-Phe-OH ≥97%

Supplier: Ambeed
Warning

A266567-1G A266567-5G
77668-150EA 47 USD
77668-150 77668-152
Ac-DL-β-Phe-OH ≥97%
Ac-DL-β-Phe-OH

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
MDL Number: MFCD00181164
CAS Number: 40638-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR