2-Fluoro-5-hydroxybenzoic acid ≥98%

Supplier: Ambeed
Warning

A266389-5G A266389-1G A266389-10G
77598-378EA 35.88 USD
77598-378 77626-390 77877-884
2-Fluoro-5-hydroxybenzoic acid ≥98%
2-Fluoro-5-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅FO₃
MDL Number: MFCD01862084
CAS Number: 51446-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR