(6-Bromohexyloxy)tert-butyldimethylsilane ≥95%

Supplier: Ambeed
Danger

Synonyms: ((6-Bromohexyl)oxy)(tert-butyl)dimethylsilane, (6-Bromohexyloxy)-tert-butyldimethylsilane

A266358-5G A266358-250MG A266358-25G
77612-552EA 121.54 USD
77612-552 77685-761 77685-762
(6-Bromohexyloxy)tert-butyldimethylsilane ≥95%
(6-Bromohexyloxy)tert-butyldimethylsilane

Precaución: for Research Use Only

Formula: C₁₂H₂₇BrOSi
MW: 295.336 g/mol
MDL Number: MFCD01863708
CAS Number: 129368-70-3
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR