Mal-PEG2-alcohol ≥97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-(2-Hydroxyethoxy)ethyl)-1H-pyrrole-2,5-dione

A266004-250MG A266004-100MG
77616-122EA 142.14 USD
77616-122 77746-456
Mal-PEG2-alcohol ≥97%
Mal-PEG2-alcohol

Precaución: for Research Use Only

Formula: C₈H₁₁NO₄
MDL Number: MFCD16780862
CAS Number: 34321-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR