6-(Benzyloxy)-1H-indole-3-carboxylic acid ≥96%

Supplier: Ambeed
Warning

A265948-100MG A265948-1G A265948-250MG A265948-5G
77872-301EA 17 USD
77872-301 77872-302 77872-303 77872-304
6-(Benzyloxy)-1H-indole-3-carboxylic acid ≥96%
6-(Benzyloxy)-1H-indole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO₃
MDL Number: MFCD17011804
CAS Number: 24370-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR