5-Bromonaphthalen-1-ol ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A265138-1G A265138-250MG
77683-810EA 242 USD
77683-810 77683-811
5-Bromonaphthalen-1-ol ≥95%, Ambeed.Inc
5-Bromonaphthalen-1-ol

Precaución: For research use only.

Formula: C₁₀H₇BrO
MW: 223.07 g/mol
MDL Number: MFCD02179789
CAS Number: 52927-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR