2-Cyanopyridine-3-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (2-Cyanopyridin-3-yl)boronic acid

A264932-100MG A264932-1G A264932-250MG A264932-5G
77872-295EA 18 USD
77872-295 77872-296 77872-297 77872-298
2-Cyanopyridine-3-boronic acid ≥98%
2-Cyanopyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BN₂O₂
MW: 147.927 g/mol
MDL Number: MFCD08458475
CAS Number: 874290-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR