2,3-Difluoropyridine-5-carboxylic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: 5,6-Difluoro-3-pyridinecarboxylic acid

A264580-1G A264580-5G A264580-250MG
77571-646EA 91.67 USD
77571-646 77672-368 77672-366
2,3-Difluoropyridine-5-carboxylic acid ≥97%
2,3-Difluoropyridine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
MDL Number: MFCD09031171
CAS Number: 851386-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR