2-(Aminomethyl)-3-fluoropyridine ≥98%

Supplier: Ambeed
Danger

A264480-250MG A264480-1G
77885-694EA 46 USD
77885-694 77885-693
2-(Aminomethyl)-3-fluoropyridine ≥98%
2-(Aminomethyl)-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MDL Number: MFCD09952741
CAS Number: 312904-51-1
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR