4-Ethynyl-1,2-difluorobenzene ≥95%

Supplier: Ambeed
Danger

Synonyms: 3,4-Difluorophenylacetylene

A264171-1G A264171-5G
77571-642EA 42.23 USD
77571-642 77571-644
4-Ethynyl-1,2-difluorobenzene ≥95%
4-Ethynyl-1,2-difluorobenzene

Precaución: for Research Use Only

Formula: C₈H₄F₂
MW: 138.12 g/mol
MDL Number: MFCD08059511
CAS Number: 143874-13-9
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR