Talabostat mesylate ≥98%

Supplier: Ambeed
Warning

Synonyms: Val-boro-pro, PT-100, Methanesulphonic acid ((R)-1-((S)-2-amino-3-methylbutanoyl)pyrrolidin-2-yl)boronic acid (1:1), (R)-1-((S)-2-amino-3-methylbutanoyl)pyrrolidin-2-ylboronic acid methanesulphonic acid

A264157-10MG A264157-1MG A264157-5MG
77612-532EA 141.82 USD
77612-532 77612-534 77612-536
Talabostat mesylate ≥98%
Talabostat mesylate

Precaución: for Research Use Only

Formula: C₁₀H₂₃BN₂OS
MDL Number: MFCD13194906
CAS Number: 150080-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR