1-Phenylprop-2-en-1-ol ≥95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with TBC

A264132-1G A264132-250MG A264132-100MG
77612-528EA 181.28 USD
77612-528 77612-530 77672-364
1-Phenylprop-2-en-1-ol ≥95% stabilized
1-Phenylprop-2-en-1-ol

Precaución: for Research Use Only

Formula: C₉H₁₀O
MW: 134.18 g/mol
MDL Number: MFCD00093987
CAS Number: 4393-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR