6-Bromo-4-chlorocinnoline ≥98%

Supplier: Ambeed
Warning

A263952-100MG A263952-1G A263952-250MG
77881-718EA 35 USD
77881-718 77881-719 77881-720
6-Bromo-4-chlorocinnoline ≥98%
6-Bromo-4-chlorocinnoline

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂
MDL Number: MFCD09834643
CAS Number: 68211-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR