Biotin C2 maleimide ≥95%

Supplier: Ambeed
Warning

Synonyms: Biotin-C2-maleimide, N-(2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide

A2638817-10MG A2638817-1MG A2638817-5MG
77598-374EA 98.4 USD
77598-374 77598-376 77877-882
Biotin C2 maleimide ≥95%
Biotin C2 maleimide

Precaución: for Research Use Only

Formula: C₁₆H₂₂SN₄O₄
MW: 366.3 g/mol
MDL Number: MFCD02094223
CAS Number: 139554-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR