1-Methylazetidin-3-one hydrochloride ≥95%

Supplier: Ambeed
Warning

A263863-100MG A263863-1G A263863-250MG
77593-636EA 29.73 USD
77593-636 77593-638 77616-114
1-Methylazetidin-3-one hydrochloride ≥95%
1-Methylazetidin-3-one hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇NO·ClH
MDL Number: MFCD29060167
CAS Number: 1818847-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR