4-((4-(2-(tert-Butyl)-5,5-dimethyl-1,3-dioxan-2-yl)benzyl)amino)-3-methoxybenzoic acid ≥98%

Supplier: Ambeed
Warning

A2638037-10MG A2638037-1MG A2638037-5MG
77626-382EA 379.04 USD
77626-382 77626-384 77626-386
4-((4-(2-(tert-Butyl)-5,5-dimethyl-1,3-dioxan-2-yl)benzyl)amino)-3-methoxybenzoic acid ≥98%
4-((4-(2-(tert-Butyl)-5,5-dimethyl-1,3-dioxan-2-yl)benzyl)amino)-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₂₅H₃₃NO₅
CAS Number: 3030588-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR