1-(4-Methylpyridin-2-yl)-3-(3-(trifluoromethyl)phenyl)thiourea ≥98%

Supplier: Ambeed
Danger

Synonyms: AA-14

A2637839-10MG A2637839-1MG A2637839-5MG
77598-368EA 100.45 USD
77598-368 77598-370 77598-372
1-(4-Methylpyridin-2-yl)-3-(3-(trifluoromethyl)phenyl)thiourea ≥98%
1-(4-Methylpyridin-2-yl)-3-(3-(trifluoromethyl)phenyl)thiourea

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₃N₃S
CAS Number: 723740-38-3
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR