4-(tert-Butoxycarbonyl)-1,4-oxazepane-2-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A263266-250MG A263266-5G
77672-356EA 86 USD
77672-356 77881-717
4-(tert-Butoxycarbonyl)-1,4-oxazepane-2-carboxylic acid ≥98%
4-(tert-Butoxycarbonyl)-1,4-oxazepane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
MDL Number: MFCD11656815
CAS Number: 1141669-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR