(R)-N-(3-(2-Chloro-5-fluorophenyl)-6-(5-cyano-[1,2,4]triazolo[1,5-a]pyridin-6-yl)-1-oxoisoindolin-4-yl)-3-fluoro-5-(trifluoromethyl)benzamide 98% ee 99.75%

Supplier: Ambeed
Warning

Synonyms: RLY-2608

A2630708-1MG A2630708-10MG
77598-366EA 500.2 USD
77598-366 77626-380
(R)-N-(3-(2-Chloro-5-fluorophenyl)-6-(5-cyano-[1,2,4]triazolo[1,5-a]pyridin-6-yl)-1-oxoisoindolin-4-yl)-3-fluoro-5-(trifluoromethyl)benzamide 98% ee 99.75%
(R)-N-(3-(2-Chloro-5-fluorophenyl)-6-(5-cyano-[1,2,4]triazolo[1,5-a]pyridin-6-yl)-1-oxoisoindolin-4-yl)-3-fluoro-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₂₉H₁₄ClF₅N₆O₂
MW: 608.91 g/mol
CAS Number: 2733573-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR