2,8-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester ≥98%

Supplier: Ambeed
Warning

A263024-100MG A263024-250MG
77872-288EA 76 USD
77872-288 77872-289
2,8-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester ≥98%
2,8-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₄H₂₆N₂O₂
MDL Number: MFCD09701315
CAS Number: 954240-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR