5-Methylimidazo[1,2-a]pyridine-2-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A262214-250MG A262214-5G
77892-497EA 15 USD
77892-497 77892-498
5-Methylimidazo[1,2-a]pyridine-2-carboxylic acid ≥98%
5-Methylimidazo[1,2-a]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C9H8N2O2
MDL Number: MFCD03419472
CAS Number: 88751-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR