(R)-2-(Methoxymethyl)-morpholine hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
A262166-250MG
A262166-100MG
77612-510EA
17.51
USD
77612-510
77885-684
(R)-2-(Methoxymethyl)-morpholine hydrochloride ≥98%
(R)-2-(Methoxymethyl)-morpholine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₂·ClH |
MDL Number:
MFCD16294689 CAS Number: 141196-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...