3-Cyano-4-fluorophenylacetic acid ≥95%

Supplier: Ambeed
Warning

A261667-250MG A261667-100MG A261667-1G
77872-284EA 33 USD
77872-284 77872-282 77872-283
3-Cyano-4-fluorophenylacetic acid ≥95%
3-Cyano-4-fluorophenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
MDL Number: MFCD03094344
CAS Number: 519059-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR