2-(5,6-Difluoro-1H-indol-3-yl)acetic acid ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A261651-100MG A261651-250MG
77683-798EA 25 USD
77683-798 77885-683
2-(5,6-Difluoro-1H-indol-3-yl)acetic acid ≥97%, Ambeed.Inc
2-(5,6-Difluoro-1H-indol-3-yl)acetic acid

Precaución: For research use only.

Formula: C₁₀H₇F₂NO₂
MDL Number: MFCD09954804
CAS Number: 126030-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR