Ethyl 3,3-dibromo-2-oxopropanoate ≥95%

Supplier: Ambeed
Warning

A261302-250MG A261302-5G
77748-493EA 25 USD
77748-493 77748-494
Ethyl 3,3-dibromo-2-oxopropanoate ≥95%
Ethyl 3,3-dibromo-2-oxopropanoate

Precaución: for Research Use Only

Formula: C₅H₆Br₂O₃
MDL Number: MFCD25967009
CAS Number: 76179-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR