3-Fluoro-2-(hydroxymethyl)-benzeneboronic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-2-(hydroxymethyl)phenyl)boronic acid, 3-Fluoro-2-(hydroxymethyl)phenylboronic acid

A261087-1G A261087-5G
77883-719EA 24 USD
77883-719 77883-720
3-Fluoro-2-(hydroxymethyl)-benzeneboronic acid ≥97%
3-Fluoro-2-(hydroxymethyl)-benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MDL Number: MFCD13181611
CAS Number: 1246633-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR