3-(2-(2-Bromoethoxy)ethoxy)propanoic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: Bromo-PEG2-C2-acid

A260571-1G A260571-100MG A260571-250MG A260571-5G
77571-608EA 121 USD
77571-608 77616-092 77616-094 77616-096
3-(2-(2-Bromoethoxy)ethoxy)propanoic acid ≥97%
3-(2-(2-Bromoethoxy)ethoxy)propanoic acid

Precaución: for Research Use Only

Formula: C₇H₁₃BrO₄
MDL Number: MFCD28122969
CAS Number: 1807503-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR