6-Chloro-4-formyl-nicotinic acid ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Chloro-4-formylnicotinic acid
A260247-1G
A260247-250MG
77612-490EA
611.82
USD
77612-490
77683-794
6-Chloro-4-formyl-nicotinic acid ≥95%
6-Chloro-4-formyl-nicotinic acid
Precaución: For research use only.
Formula:
C₇H₄ClNO₃ MW: 185.565 g/mol |
MDL Number:
MFCD18258739 CAS Number: 1031433-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...