tert-Butyl (S)-2-methyl-3-oxoazetidine-1-carboxylate ≥97%

Supplier: Ambeed
Danger

A259710-1G A259710-250MG A259710-5G
77748-490EA 730 USD
77748-490 77748-491 77748-492
tert-Butyl (S)-2-methyl-3-oxoazetidine-1-carboxylate ≥97%
tert-Butyl (S)-2-methyl-3-oxoazetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
MDL Number: MFCD28501893
CAS Number: 171919-76-9
UN: 3335
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR