Minocycline hydrochloride ≥99%

Supplier: Ambeed
Warning

Synonyms: (4S,4aS,5aR,12aS)-4,7-Bis(dimethylamino)-3,10,12,12a-tetrahydroxy-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride, Minocycline HCl

A259020-1G A259020-100MG A259020-250MG A259020-25G A259020-5G A259020-100G
77593-616EA 20.5 USD
77593-616 77601-088 77601-090 77601-092 77601-094 77668-112
Minocycline hydrochloride ≥99%
Minocycline hydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₂₇N₃O₇
MDL Number: MFCD00083669
CAS Number: 13614-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR