N-(6-Chloro-3-((4,5-dihydro-1H-imidazol-2-yl)methoxy)-2-methylphenyl)methanesulfonamide ≥97%

Supplier: Ambeed
Warning

Synonyms: Dabuzalgron

A258735-1MG A258735-10MG A258735-5MG
77616-086EA 28.84 USD
77616-086 77746-422 77746-423
N-(6-Chloro-3-((4,5-dihydro-1H-imidazol-2-yl)methoxy)-2-methylphenyl)methanesulfonamide ≥97%
N-(6-Chloro-3-((4,5-dihydro-1H-imidazol-2-yl)methoxy)-2-methylphenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆ClN₃O₃S
MW: 317.79 g/mol
MDL Number: MFCD13195546
CAS Number: 219311-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR