5-(Trifluoromethyl)isoxazol-3-amine ≥98%

Supplier: Ambeed
Warning

A258366-100MG A258366-250MG
77612-476EA 99.91 USD
77612-476 77683-787
5-(Trifluoromethyl)isoxazol-3-amine ≥98%
5-(Trifluoromethyl)isoxazol-3-amine

Precaución: For research use only.

Formula: C₄H₃F₃N₂O
MW: 152.07 g/mol
MDL Number: MFCD11847205
CAS Number: 110234-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR